Learn More
MSC Mitutoyo Electronic Drop Indicator, 2 in Max, +/-.0002 in Accuracy, Digital, Straight
Catalog No. 5000512649
Encompass
This electronic drop indicator provides precise digital measurements with an intuitive analog bar for easy reading of origin and tolerance values. It is equipped with a data output port, allowing integration into measurement networking and statistical process control systems.
  • Straight connection type
  • Accuracy of +/-.0002 in
  • Measuring force of 2.30 newtons
  • Maximum measurement capacity of 2 in
  • Flat back type
  • Digital display type
  • Long form calibration certificate included
  • Carbide contact point material
  • Battery-powered
  • Includes one SR44 silver oxide battery
  • Multiple resolution options: 0.00002, 0.00005, 0.0001, 0.0002, 0.0005 in
Catalog No. 50-005-12649 Supplier MSC Supplier No. 76061712
This electronic drop indicator provides precise digital measurements with an intuitive analog bar for easy reading of origin and tolerance values. It is equipped with a data output port, allowing integration into measurement networking and statistical process control systems.
  • Straight connection type
  • Accuracy of +/-.0002 in
  • Measuring force of 2.30 newtons
  • Maximum measurement capacity of 2 in
  • Flat back type
  • Digital display type
  • Long form calibration certificate included
  • Carbide contact point material
  • Battery-powered
  • Includes one SR44 silver oxide battery
  • Multiple resolution options: 0.00002, 0.00005, 0.0001, 0.0002, 0.0005 in