Learn More
MSC Mitutoyo Electronic Inside Micrometer, 1.2000-1.6000 in, 0.00005 in Resolution, IP65
Catalog No. 5000528606
Encompass
Mitutoyo Digimatic Holtest internal micrometers provide precise internal measurements. Their titanium-coated measuring pins offer durability and impact resistance, allowing accurate readings in blind holes. A three-point design ensures self-centering for stable measurements.
  • Titanium-coated measuring pins for durability, impact resistance, and accurate blind hole readings.
  • Three-point design ensures self-centering for stable measurements (above 8 mm bore diameter).
  • Large LCD readout and digital display for easy readability.
  • Advanced functions: presetting, power on/off, data hold, data output, and alarms.
  • Ratchet stop for constant force measurement.
  • ABS and INC measuring modes for efficiency.
  • Statistical process control and measurement support system compatibility.
  • Includes plastic case; IP65 rated; TUV certified.
Catalog No. 50-005-28606 Supplier MSC Supplier No. 51589828
Mitutoyo Digimatic Holtest internal micrometers provide precise internal measurements. Their titanium-coated measuring pins offer durability and impact resistance, allowing accurate readings in blind holes. A three-point design ensures self-centering for stable measurements.
  • Titanium-coated measuring pins for durability, impact resistance, and accurate blind hole readings.
  • Three-point design ensures self-centering for stable measurements (above 8 mm bore diameter).
  • Large LCD readout and digital display for easy readability.
  • Advanced functions: presetting, power on/off, data hold, data output, and alarms.
  • Ratchet stop for constant force measurement.
  • ABS and INC measuring modes for efficiency.
  • Statistical process control and measurement support system compatibility.
  • Includes plastic case; IP65 rated; TUV certified.