Learn More
MSC Mitutoyo Electronic Inside Micrometer, 8-40 in, 0.0001 in Resolution, IP65
Catalog No. 5000534948
Encompass
This electronic inside micrometer delivers precise internal diameter measurements with a digital display for easy reading. Engineered for durability, it features carbide-tipped measuring faces and an IP65 protection rating, making it suitable for demanding industrial and laboratory environments.
  • Measures a wide range of internal diameters, utilizing extension pipes and anvils.
  • Offers a high resolution of 0.0001 inch and an accuracy of 0.00015 "
  • IP65 rated for excellent protection against dust and water ingress.
  • Equipped with durable, carbide-tipped measuring faces.
  • Provides data output for integration with external systems.
  • Includes 1, 2, 4, 8, and 16 inch extension pipes, supplied in a wooden case.
  • Features Zero/ABS, Data hold, Preset, and inch/mm conversion functions.
  • Powered by an SR44 battery with an approximate 8-month life.
Catalog No. 50-005-34948 Supplier MSC Supplier No. 47860283
This electronic inside micrometer delivers precise internal diameter measurements with a digital display for easy reading. Engineered for durability, it features carbide-tipped measuring faces and an IP65 protection rating, making it suitable for demanding industrial and laboratory environments.
  • Measures a wide range of internal diameters, utilizing extension pipes and anvils.
  • Offers a high resolution of 0.0001 inch and an accuracy of 0.00015 "
  • IP65 rated for excellent protection against dust and water ingress.
  • Equipped with durable, carbide-tipped measuring faces.
  • Provides data output for integration with external systems.
  • Includes 1, 2, 4, 8, and 16 inch extension pipes, supplied in a wooden case.
  • Features Zero/ABS, Data hold, Preset, and inch/mm conversion functions.
  • Powered by an SR44 battery with an approximate 8-month life.