Learn More
MSC Mitutoyo Electronic Interchangeable Anvil Micrometer, 0 to 6 inch, IP65, 6 Anvils
Catalog No. 5000515296
Encompass
This electronic interchangeable anvil micrometer provides precise measurements across a 0 to 6 inch (0 to 152.4 mm) range. Designed with a ratchet stop thimble and an IP65 ingress protection rating, it offers reliable performance and durability in challenging shop floor environments, helping to reduce measurement errors.
  • Electronic operation
  • Ratchet stop thimble
  • Data output capability
  • Includes one SR44 battery
  • IP65 ingress protection rating (waterproof against coolant and dirt)
  • SPC output for data collection
  • Carbide tipped measuring faces
  • Comes with 6 anvils and 5 standards
  • Compatible with common SPC systems
  • Features ABS/INC, origin set, inch/metric conversion, and zero setting
  • Locking clamp for stable measurements
  • Helps eliminate micrometer reading errors
Catalog No. 50-005-15296 Supplier MSC Supplier No. 92293935
This electronic interchangeable anvil micrometer provides precise measurements across a 0 to 6 inch (0 to 152.4 mm) range. Designed with a ratchet stop thimble and an IP65 ingress protection rating, it offers reliable performance and durability in challenging shop floor environments, helping to reduce measurement errors.
  • Electronic operation
  • Ratchet stop thimble
  • Data output capability
  • Includes one SR44 battery
  • IP65 ingress protection rating (waterproof against coolant and dirt)
  • SPC output for data collection
  • Carbide tipped measuring faces
  • Comes with 6 anvils and 5 standards
  • Compatible with common SPC systems
  • Features ABS/INC, origin set, inch/metric conversion, and zero setting
  • Locking clamp for stable measurements
  • Helps eliminate micrometer reading errors