Learn More
MSC Mitutoyo Electronic Micrometer Head, 0 to 25.4mm, 6.35mm Flat Spindle, Clamp Nut
Catalog No. 5000534640
Encompass
This electronic micrometer head provides precise measurements from 0 to 25.4mm. It features a 6.35mm flat carbide spindle and a clear digital display for accurate readings, making it suitable for various laboratory and industrial applications.
  • Measurement values are clearly displayed in 0.001 mm increments
  • Resolution of 0.00005 in
  • Accuracy of 0.0001 in
  • Measuring face made of carbide
  • Rotating spindle for ease of use
  • Supplied with a clamp nut
  • Data output capability
  • Includes battery SR-44
Catalog No. 50-005-34640 Supplier MSC Supplier No. 75638510
This electronic micrometer head provides precise measurements from 0 to 25.4mm. It features a 6.35mm flat carbide spindle and a clear digital display for accurate readings, making it suitable for various laboratory and industrial applications.
  • Measurement values are clearly displayed in 0.001 mm increments
  • Resolution of 0.00005 in
  • Accuracy of 0.0001 in
  • Measuring face made of carbide
  • Rotating spindle for ease of use
  • Supplied with a clamp nut
  • Data output capability
  • Includes battery SR-44