Learn More
MSC Mitutoyo Electronic Outside Micrometer, 0.00 mm to 1 in, Solid Carbide Face
Catalog No. 5000510294
Encompass
This electronic outside micrometer is designed for precise measurements, featuring a deep frame for extended reach. It offers high accuracy and resolution, making it suitable for demanding laboratory and industrial applications.
  • High resolution for precise measurements.
  • Accurate readings for critical applications.
  • Deep throat design allows for extended reach.
  • Features solid carbide measuring faces for durability.
  • Equipped with a ratchet stop thimble for consistent force.
  • Includes a plastic case for protection and storage.
  • IP65 rated for protection against dust and water jets.
  • Powered by a single SR44 battery (included).
Catalog No. 50-005-10294 Supplier MSC Supplier No. 86475472
This electronic outside micrometer is designed for precise measurements, featuring a deep frame for extended reach. It offers high accuracy and resolution, making it suitable for demanding laboratory and industrial applications.
  • High resolution for precise measurements.
  • Accurate readings for critical applications.
  • Deep throat design allows for extended reach.
  • Features solid carbide measuring faces for durability.
  • Equipped with a ratchet stop thimble for consistent force.
  • Includes a plastic case for protection and storage.
  • IP65 rated for protection against dust and water jets.
  • Powered by a single SR44 battery (included).