Learn More
MSC Mitutoyo Electronic Outside Micrometer, 0-1 in, 0.00005 in Resolution, IP65
Supplier:  MSC 59017848
Encompass
This electronic outside micrometer is a standard coolant-proof digital micrometer designed for precise measurements. It offers a 0-1 inch range, 0.00005 inch resolution, and an IP65 rating, making it suitable for various demanding environments.
  • Outstanding accuracy
  • Highly environmentally resistant
  • Ratchet thimble provides better operability for one-handed use
  • Includes battery, box, setting standard (from 1 inch upward), certificate of inspection, and key
  • Approximately 2.4-year battery life reduces frequent battery replacement
  • Nonslip surface with raised dots on frame cover and surface panel for stable handhold measurement
  • Ratchet thimble mechanism ensures repeatable results with constant measuring force
  • Oilproof materials used in all plastic components
Catalog No. 50-005-30146