Learn More
MSC Mitutoyo Electronic Outside Micrometer, 0-1 in, Solid Carbide Face, IP65
Catalog No. 5000533565
Encompass
This electronic outside micrometer features a measurement range of 0.00 mm to 1 in with a solid carbide face and an IP65 rating. It offers a resolution of 0.000050 in and an accuracy of ±0.000050 in / ±0.0010 mm. The thimble type is friction and ratchet.
  • Outstanding accuracy
  • Includes certification of inspection
  • Includes plastic case
  • Proprietary inspection certificate standards
Catalog No. 50-005-33565 Supplier MSC Supplier No. 70538905
This electronic outside micrometer features a measurement range of 0.00 mm to 1 in with a solid carbide face and an IP65 rating. It offers a resolution of 0.000050 in and an accuracy of ±0.000050 in / ±0.0010 mm. The thimble type is friction and ratchet.
  • Outstanding accuracy
  • Includes certification of inspection
  • Includes plastic case
  • Proprietary inspection certificate standards