Learn More
MSC Mitutoyo Electronic Test Indicator, 0.001000 in Accuracy, Solid Carbide Contact
Catalog No. 5000512415
Encompass
This electronic test indicator provides precise measurements with an accuracy of 0.001000 in and features a durable solid carbide contact point. It is powered by a single SR44 battery, which is included.
  • Accuracy of 0.001000 in
  • Solid carbide contact point
  • Contact point length is 8.7 mm
  • Measures from 0 to 1 decimal inch
  • Resolution of 0.0010
  • Powered by a single SR44 battery
  • Battery is included
  • Complies with ANSI standards
  • Part of the ID-C ABSOLUTE Digimatic Indicator series
  • Manufactured in Japan
Catalog No. 50-005-12415 Supplier MSC Supplier No. 03723863
This electronic test indicator provides precise measurements with an accuracy of 0.001000 in and features a durable solid carbide contact point. It is powered by a single SR44 battery, which is included.
  • Accuracy of 0.001000 in
  • Solid carbide contact point
  • Contact point length is 8.7 mm
  • Measures from 0 to 1 decimal inch
  • Resolution of 0.0010
  • Powered by a single SR44 battery
  • Battery is included
  • Complies with ANSI standards
  • Part of the ID-C ABSOLUTE Digimatic Indicator series
  • Manufactured in Japan