Learn More
MSC Mitutoyo Electronic V-Anvil Micrometer, 0.05 to 0.6 in, 4 μm, 3 Flutes
Catalog No. 5000530576
Encompass
This electronic V-Anvil micrometer is designed for precise measurements of 3-flute workpieces. It offers a measurement range of 0.05 to 0.6 inches with an accuracy of 4 μm. Featuring a ratchet stop thimble, this tool ensures consistent measurement force, and its carbide anvil and spindle provide durability.
  • Electronic operation for precise readings.
  • Ratchet thimble for consistent measurement force.
  • Measures 3-flute workpieces.
  • High resolution of 0.0001 "
  • Durable carbide anvil and spindle materials.
  • Equipped with data output capability.
  • Includes setting standard, battery, box, and key.
Catalog No. 50-005-30576 Supplier MSC Supplier No. 72076342
This electronic V-Anvil micrometer is designed for precise measurements of 3-flute workpieces. It offers a measurement range of 0.05 to 0.6 inches with an accuracy of 4 μm. Featuring a ratchet stop thimble, this tool ensures consistent measurement force, and its carbide anvil and spindle provide durability.
  • Electronic operation for precise readings.
  • Ratchet thimble for consistent measurement force.
  • Measures 3-flute workpieces.
  • High resolution of 0.0001 "
  • Durable carbide anvil and spindle materials.
  • Equipped with data output capability.
  • Includes setting standard, battery, box, and key.