Learn More
MSC Mitutoyo Extension Rod, 500 mm, M26x1.5 Thread
Catalog No. 5000529565
Encompass
This extension rod is designed for bore gages, providing an extended reach for measurement applications. It is compatible with specific series of bore gages, making it suitable for various precision measuring tasks.
  • Extension length of 500 mm
  • Compatible with bore gage series 511-521 and 511-522
  • Diameter of 28 mm
  • Features M26x1.5 thread
Catalog No. 50-005-29565 Supplier MSC Supplier No. 08965253
This extension rod is designed for bore gages, providing an extended reach for measurement applications. It is compatible with specific series of bore gages, making it suitable for various precision measuring tasks.
  • Extension length of 500 mm
  • Compatible with bore gage series 511-521 and 511-522
  • Diameter of 28 mm
  • Features M26x1.5 thread