Learn More
MSC Mitutoyo Extension Rod, For Precision Measuring Tools
Catalog No. 5000533242
Encompass
This extension rod is designed for use with precision measuring tools and instruments, including drop indicators. It functions as an accessory to enhance the functionality and reach of compatible measurement devices.
  • Accessory type: Extension rod
  • For use with precision measuring tools/instruments
  • Not calibrated
  • No traceability certification included
  • Manufactured in Japan
Catalog No. 50-005-33242 Supplier MSC Supplier No. 04241113
This extension rod is designed for use with precision measuring tools and instruments, including drop indicators. It functions as an accessory to enhance the functionality and reach of compatible measurement devices.
  • Accessory type: Extension rod
  • For use with precision measuring tools/instruments
  • Not calibrated
  • No traceability certification included
  • Manufactured in Japan