Learn More
MSC Mitutoyo Gage Block Accessory Set, Rectangular, 14 Piece
Catalog No. 5000529852
Encompass
This accessory set is designed for rectangular gage blocks, expanding their applications and facilitating the easy and quick assembly of precision measuring instruments. It is suitable for use with both steel and CERA rectangular gage blocks.
  • Assortment of accessories for gage blocks
  • Includes multiple holders of varying lengths
  • Features half round jaws in different sizes
  • Comes with scriber point, center point, and triangular straightness edge
  • Total of 14 pieces in the set
  • Compatible with steel and CERA rectangular gage blocks
Catalog No. 50-005-29852 Supplier MSC Supplier No. 06250955
This accessory set is designed for rectangular gage blocks, expanding their applications and facilitating the easy and quick assembly of precision measuring instruments. It is suitable for use with both steel and CERA rectangular gage blocks.
  • Assortment of accessories for gage blocks
  • Includes multiple holders of varying lengths
  • Features half round jaws in different sizes
  • Comes with scriber point, center point, and triangular straightness edge
  • Total of 14 pieces in the set
  • Compatible with steel and CERA rectangular gage blocks