Learn More
MSC Mitutoyo Gage Block, Rectangle, 0.101 in, Steel, Grade AS-1
Catalog No. 5000529180
Encompass
This is a Rectangle Steel Gage Block, featuring a precise 0.101 in dimension and an AS-1 accuracy grade. It is suitable for precision measurement and calibration tasks, manufactured from steel and adhering to ASME standards.
  • Rectangle shape
  • Accuracy grade AS-1
  • Material: Steel
  • Size: 0.101 in
  • Includes Certificate of Inspection
  • Adheres to ASME standards
Catalog No. 50-005-29180 Supplier MSC Supplier No. 04069209
This is a Rectangle Steel Gage Block, featuring a precise 0.101 in dimension and an AS-1 accuracy grade. It is suitable for precision measurement and calibration tasks, manufactured from steel and adhering to ASME standards.
  • Rectangle shape
  • Accuracy grade AS-1
  • Material: Steel
  • Size: 0.101 in
  • Includes Certificate of Inspection
  • Adheres to ASME standards