Learn More
MSC Mitutoyo Gage Block, Rectangle, 0.109 in, AS-1 Grade, ASME Standard
Catalog No. 5000513125
Encompass
This Mitutoyo Rectangle Steel Gage Block, with a size of 0.109 inches and an AS-1 accuracy grade, is manufactured to ASME standards. It is supplied with a certificate of inspection, ensuring precision for various measurement applications.
  • Rectangle shape.
  • Accuracy grade AS-1.
  • Gage block type.
  • Constructed from steel.
  • Size is 0.109 inches.
  • Meets ASME standards.
  • Includes certificate of inspection.
Catalog No. 50-005-13125 Supplier MSC Supplier No. 01135193
This Mitutoyo Rectangle Steel Gage Block, with a size of 0.109 inches and an AS-1 accuracy grade, is manufactured to ASME standards. It is supplied with a certificate of inspection, ensuring precision for various measurement applications.
  • Rectangle shape.
  • Accuracy grade AS-1.
  • Gage block type.
  • Constructed from steel.
  • Size is 0.109 inches.
  • Meets ASME standards.
  • Includes certificate of inspection.