Learn More
MSC Mitutoyo Gage Block, Rectangle, 0.115 in, Steel, AS-1 Grade
Catalog No. 5000514221
Encompass
This precision gage block is crafted from durable steel, featuring a rectangular shape and an AS-1 accuracy grade. It is manufactured to meet ASME standards, making it suitable for a wide range of calibration and measurement applications requiring high accuracy.
  • Rectangle shape for precise fit
  • AS-1 accuracy grade ensures high precision
  • Constructed from steel for durability
  • Measures 0.115 inch in length
  • Complies with ASME standards for reliability
  • Includes a certificate of inspection
Catalog No. 50-005-14221 Supplier MSC Supplier No. 88609128
This precision gage block is crafted from durable steel, featuring a rectangular shape and an AS-1 accuracy grade. It is manufactured to meet ASME standards, making it suitable for a wide range of calibration and measurement applications requiring high accuracy.
  • Rectangle shape for precise fit
  • AS-1 accuracy grade ensures high precision
  • Constructed from steel for durability
  • Measures 0.115 inch in length
  • Complies with ASME standards for reliability
  • Includes a certificate of inspection