Learn More
MSC Mitutoyo Gage Block, Rectangle, 0.126 in, AS-1, High-Carbon/High-Chrome Steel
Catalog No. 5000515593
Encompass
This rectangular gage block is designed for precise measurements. It is constructed from high-carbon, high-chrome steel, offering excellent hardness and wear resistance for long-lasting performance.
  • Identification number etched in block for traceability.
  • Rectangular base for accurate wringing.
  • High-carbon, high-chrome steel provides high hardness and wear resistance.
Catalog No. 50-005-15593 Supplier MSC Supplier No. 88609185
This rectangular gage block is designed for precise measurements. It is constructed from high-carbon, high-chrome steel, offering excellent hardness and wear resistance for long-lasting performance.
  • Identification number etched in block for traceability.
  • Rectangular base for accurate wringing.
  • High-carbon, high-chrome steel provides high hardness and wear resistance.