Learn More
MSC Mitutoyo Gage Block, Rectangle, 0.132 in, Grade AS-1, Steel
Catalog No. 5000534443
Encompass
This gage block is a rectangular steel block with a precise length of 0.132 inches and an AS-1 accuracy grade. It is manufactured to ASME standards and includes a certificate of inspection, making it suitable for applications requiring high precision.
  • Rectangular shape
  • AS-1 accuracy grade
  • Constructed from steel
  • Overall length of 0.132 inches
  • Complies with ASME standards
  • Includes a certificate of inspection
Catalog No. 50-005-34443 Supplier MSC Supplier No. 88609243
This gage block is a rectangular steel block with a precise length of 0.132 inches and an AS-1 accuracy grade. It is manufactured to ASME standards and includes a certificate of inspection, making it suitable for applications requiring high precision.
  • Rectangular shape
  • AS-1 accuracy grade
  • Constructed from steel
  • Overall length of 0.132 inches
  • Complies with ASME standards
  • Includes a certificate of inspection