Learn More
MSC Mitutoyo Gage Block, Rectangle, Steel, 0.104 in, Grade AS-1
Catalog No. 5000529530
Encompass
This rectangle steel gage block is designed for precise measurement applications, offering a high level of accuracy and durability. It has a nominal size of 0.104 inches and adheres to ASME standards, making it suitable for a variety of industrial and scientific settings.
  • Rectangle shape
  • AS-1 accuracy grade
  • Constructed from durable steel
  • 0.104 inch size
  • Meets ASME standards
  • Includes a certificate of inspection
  • Country of origin: Japan
Catalog No. 50-005-29530 Supplier MSC Supplier No. 06539795
This rectangle steel gage block is designed for precise measurement applications, offering a high level of accuracy and durability. It has a nominal size of 0.104 inches and adheres to ASME standards, making it suitable for a variety of industrial and scientific settings.
  • Rectangle shape
  • AS-1 accuracy grade
  • Constructed from durable steel
  • 0.104 inch size
  • Meets ASME standards
  • Includes a certificate of inspection
  • Country of origin: Japan