Learn More
MSC Mitutoyo Gage Block, Rectangle, Steel, 0.105 in, Grade 0
Catalog No. 5000527504
Encompass
This precision rectangle steel gage block offers an accuracy grade of 0, making it ideal for applications requiring high precision measurement. Its robust construction and adherence to standards ensure reliable performance in various industrial and laboratory settings.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Rectangle shape
  • Steel material
  • Size of 0.105 in
  • Meets ASME standards
Catalog No. 50-005-27504 Supplier MSC Supplier No. 59554022
This precision rectangle steel gage block offers an accuracy grade of 0, making it ideal for applications requiring high precision measurement. Its robust construction and adherence to standards ensure reliable performance in various industrial and laboratory settings.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Rectangle shape
  • Steel material
  • Size of 0.105 in
  • Meets ASME standards