Learn More
MSC Mitutoyo Gage Block, Rectangle, Steel, 0.126 inch, Grade 0
Catalog No. 5000534520
Encompass
This gage block is a precision measuring tool designed for accurate inspection and calibration. Made from durable steel, it features a rectangular shape and a Grade 0 accuracy for reliable performance in various applications.
  • Includes Certificate of Inspection
  • Rectangular shape
  • Grade 0 accuracy
  • Made of steel
  • 0.126 inch size
  • Meets ASME standards
Catalog No. 50-005-34520 Supplier MSC Supplier No. 86668654
This gage block is a precision measuring tool designed for accurate inspection and calibration. Made from durable steel, it features a rectangular shape and a Grade 0 accuracy for reliable performance in various applications.
  • Includes Certificate of Inspection
  • Rectangular shape
  • Grade 0 accuracy
  • Made of steel
  • 0.126 inch size
  • Meets ASME standards