Learn More
MSC Mitutoyo Gage Block, Rectangle, Steel, 0.4 in, AS-1
Catalog No. 5000516448
Encompass
This Rectangle Steel Gage Block provides precise measurement for calibration and inspection tasks. It features a 0.4 inch size and an accuracy grade of AS-1, making it suitable for demanding applications requiring high precision.
  • Rectangle shape for versatile use
  • Constructed from durable steel
  • 0.4 inch size for precise measurements
  • AS-1 accuracy grade ensures high precision
  • Includes a Certificate of Inspection for verified accuracy
  • Manufactured to ASME standards
Catalog No. 50-005-16448 Supplier MSC Supplier No. 40455354
This Rectangle Steel Gage Block provides precise measurement for calibration and inspection tasks. It features a 0.4 inch size and an accuracy grade of AS-1, making it suitable for demanding applications requiring high precision.
  • Rectangle shape for versatile use
  • Constructed from durable steel
  • 0.4 inch size for precise measurements
  • AS-1 accuracy grade ensures high precision
  • Includes a Certificate of Inspection for verified accuracy
  • Manufactured to ASME standards