Learn More
MSC Mitutoyo Gage Block, Rectangle, Steel, 0.9 Inch, Grade AS-1
Catalog No. 5000534738
Encompass
This precision gage block is designed for accurate measurement in various industrial and laboratory applications. Made from durable steel and conforming to ASME standards, it provides reliable performance for calibration and inspection tasks.
  • Rectangle shape for versatile use.
  • Constructed from high-quality steel for durability.
  • Precisely sized at 0.90000 decimal "
  • Features AS-1 accuracy grade for reliable measurements.
  • Includes a certificate of inspection for quality assurance.
  • Conforms to ASME standards.
  • Solid construction without through holes.
Catalog No. 50-005-34738 Supplier MSC Supplier No. 74446410
This precision gage block is designed for accurate measurement in various industrial and laboratory applications. Made from durable steel and conforming to ASME standards, it provides reliable performance for calibration and inspection tasks.
  • Rectangle shape for versatile use.
  • Constructed from high-quality steel for durability.
  • Precisely sized at 0.90000 decimal "
  • Features AS-1 accuracy grade for reliable measurements.
  • Includes a certificate of inspection for quality assurance.
  • Conforms to ASME standards.
  • Solid construction without through holes.