Learn More
MSC Mitutoyo Gage Block, Rectangular, Steel, 7 in, Grade AS-1
Catalog No. 5000510260
Encompass
Mitutoyo gage blocks are primary standards for precise dimensional quality control in manufacturing. This individual 7-inch rectangular steel gage block, Grade AS-1, is ideal for applications requiring frequent use of a specific length and comes with an Inspection Certificate.
  • Precision standards for dimensional quality control.
  • High accuracy with traceability.
  • Advanced lapping for excellent flatness and surface roughness.
  • Made from high-carbon, high-chrome steel.
  • Repeated heat treatment reduces deterioration over time.
  • Supplied with an inspection certificate.
  • Ideal for repeated use of a single length.
  • Superior surface qualities and corrosion resistance.
  • Laser-inscribed black characters are clearly visible.
Catalog No. 50-005-10260 Supplier MSC Supplier No. 88609425
Mitutoyo gage blocks are primary standards for precise dimensional quality control in manufacturing. This individual 7-inch rectangular steel gage block, Grade AS-1, is ideal for applications requiring frequent use of a specific length and comes with an Inspection Certificate.
  • Precision standards for dimensional quality control.
  • High accuracy with traceability.
  • Advanced lapping for excellent flatness and surface roughness.
  • Made from high-carbon, high-chrome steel.
  • Repeated heat treatment reduces deterioration over time.
  • Supplied with an inspection certificate.
  • Ideal for repeated use of a single length.
  • Superior surface qualities and corrosion resistance.
  • Laser-inscribed black characters are clearly visible.