Precision gage blocks are essential primary standards for dimensional quality control in manufacturing. This Mitutoyo gage block set ensures high accuracy and reliability. Comprising 81 pieces, this set is primarily made of CERA (ceramic) blocks, with the 2, 3, and 4 inch blocks made of steel. Note that while the manufacturer's catalog specifies CERA as the primary material, the distributor lists the material simply as steel. The set achieves AS-2 Grade accuracy and is designed to meet or exceed ASME B89.1.9-2002 specifications.
- High accuracy for dimensional quality control
- Includes 81 blocks
- Primarily CERA (ceramic) material for superior surface quality
- Corrosion and abrasion resistance, dimensional stability
- Meets or exceeds ASME B89.1.9-2002 specifications
- Includes a wooden case and inspection certificate
- Individually serialized for traceability