This high-precision gage block set is designed for accurate dimensional measurements. This 36-piece set features square, inch-sized blocks crafted from durable steel and manufactured to AS-1 grade standards. Each set includes an inspection certificate, assuring quality and traceability, and the gage blocks are engineered for stable orientation, suitable for various demanding metrology applications.
- 36 blocks included per set.
- Manufactured to AS-1 grade for high accuracy.
- Constructed from high-carbon, high-chrome steel for durability.
- Designed for stable longitudinal and lateral orientation.
- Provided with an inspection certificate.
- Suitable for cutting tool positioning and angle measurement.
- Utilizes advanced lapping techniques for superior wringing.
- Meets or exceeds specifications for flatness, parallelism, and surface finish.