Learn More
MSC Mitutoyo Gage Block, Square, 0.11 in, Steel, Grade 0
Catalog No. 5000531740
Encompass
This gage block is a square steel block with an accuracy grade of 0, ideal for precise measurement applications. It comes with a certificate of inspection, ensuring its quality and adherence to specifications.
  • Square shape
  • Accuracy grade 0
  • Overall length of 0.11 in
  • Constructed from steel
  • Meets ASME standards
  • Includes certificate of inspection
Catalog No. 50-005-31740 Supplier MSC Supplier No. 63288864
This gage block is a square steel block with an accuracy grade of 0, ideal for precise measurement applications. It comes with a certificate of inspection, ensuring its quality and adherence to specifications.
  • Square shape
  • Accuracy grade 0
  • Overall length of 0.11 in
  • Constructed from steel
  • Meets ASME standards
  • Includes certificate of inspection