Learn More
MSC Mitutoyo Gage Block, Square, 0.121 in, Steel, Grade 0
Catalog No. 5000534653
Encompass
This square steel gage block measures 0.121 in and features an Accuracy Grade of 0, ensuring high precision for calibration, layout, and machine setup tasks. It is made from durable steel and conforms to ASME standards. Each gage block includes a Certificate of Inspection, guaranteeing its quality and accuracy.
  • Accuracy Grade 0
  • Includes Certificate of Inspection
  • Square shape
  • 0.121 in size
  • Steel material
  • Conforms to ASME standards
Catalog No. 50-005-34653 Supplier MSC Supplier No. 88609581
This square steel gage block measures 0.121 in and features an Accuracy Grade of 0, ensuring high precision for calibration, layout, and machine setup tasks. It is made from durable steel and conforms to ASME standards. Each gage block includes a Certificate of Inspection, guaranteeing its quality and accuracy.
  • Accuracy Grade 0
  • Includes Certificate of Inspection
  • Square shape
  • 0.121 in size
  • Steel material
  • Conforms to ASME standards