Learn More
MSC Mitutoyo Gage Block, Square, 0.13 in, Grade 0, Steel
Catalog No. 5000531142
Encompass
This Mitutoyo gage block is a precision measuring tool designed for high-accuracy applications. It features a square shape and is made from steel, ensuring durability and stability during use. With an accuracy grade of 0 and a size of 0.13 inch, it is suitable for demanding calibration and inspection tasks.
  • Includes a certificate of inspection
  • Square shape for stable measurements
  • Constructed from durable steel
  • Grade 0 accuracy for high-precision applications
  • Designed for calibration and inspection tasks
Catalog No. 50-005-31142 Supplier MSC Supplier No. 88609607
This Mitutoyo gage block is a precision measuring tool designed for high-accuracy applications. It features a square shape and is made from steel, ensuring durability and stability during use. With an accuracy grade of 0 and a size of 0.13 inch, it is suitable for demanding calibration and inspection tasks.
  • Includes a certificate of inspection
  • Square shape for stable measurements
  • Constructed from durable steel
  • Grade 0 accuracy for high-precision applications
  • Designed for calibration and inspection tasks