Learn More
MSC Mitutoyo Gage Block, Square, 0.136 in, Steel, Grade 0
Catalog No. 5000534490
Encompass
This square steel gage block is designed for precision measurements, featuring a Grade 0 accuracy. It includes a certificate of inspection, ensuring its quality and reliability for demanding applications.
  • Square shape
  • Accuracy grade 0
  • Made from steel
  • Size of 0.136 in
  • Conforms to ASME standards
  • Includes certificate of inspection
Catalog No. 50-005-34490 Supplier MSC Supplier No. 88609631
This square steel gage block is designed for precision measurements, featuring a Grade 0 accuracy. It includes a certificate of inspection, ensuring its quality and reliability for demanding applications.
  • Square shape
  • Accuracy grade 0
  • Made from steel
  • Size of 0.136 in
  • Conforms to ASME standards
  • Includes certificate of inspection