Learn More
MSC Mitutoyo Gage Block, Square, 0.75 in, Steel, ASME Grade 0
Catalog No. 5000528095
Encompass
This square gage block is crafted from high-carbon, high-chrome steel, ensuring exceptional hardness and durability. It is an ASME Grade 0 block, specifically designed for use in inspection areas to verify the accuracy of other gages and for setting electronic measuring devices. Each block is supplied with a Certificate of Inspection.
  • Includes a certificate of inspection
  • Made from high-carbon, high-chrome steel
  • Suitable for verifying other gages
  • Ideal for setting electronic measuring devices
  • Square shape for stable orientation
  • Meets ASME standards
Catalog No. 50-005-28095 Supplier MSC Supplier No. 63288880
This square gage block is crafted from high-carbon, high-chrome steel, ensuring exceptional hardness and durability. It is an ASME Grade 0 block, specifically designed for use in inspection areas to verify the accuracy of other gages and for setting electronic measuring devices. Each block is supplied with a Certificate of Inspection.
  • Includes a certificate of inspection
  • Made from high-carbon, high-chrome steel
  • Suitable for verifying other gages
  • Ideal for setting electronic measuring devices
  • Square shape for stable orientation
  • Meets ASME standards