Learn More
MSC Mitutoyo Gage Block, Square, 0.8 in, Steel, Grade 0
Catalog No. 5000535095
Encompass
This square steel gage block is a precise measuring instrument designed for accuracy and durability. It features a square shape and is made from high-quality steel, ensuring stability and long-term performance. This Grade 0 gage block is ideal for calibrating other measuring tools and maintaining measurement accuracy in various industrial and laboratory settings.
  • Square shape for stable positioning during measurements.
  • Grade 0 accuracy ensures high precision for calibration and inspection tasks.
  • Made from durable steel for long-lasting performance.
  • 0.8 in size suitable for various measurement applications.
  • Conforms to ASME standards for reliable quality.
  • Includes a certificate of inspection for traceability and quality assurance.
  • Manufactured in Japan, known for precision engineering.
Catalog No. 50-005-35095 Supplier MSC Supplier No. 44558260
This square steel gage block is a precise measuring instrument designed for accuracy and durability. It features a square shape and is made from high-quality steel, ensuring stability and long-term performance. This Grade 0 gage block is ideal for calibrating other measuring tools and maintaining measurement accuracy in various industrial and laboratory settings.
  • Square shape for stable positioning during measurements.
  • Grade 0 accuracy ensures high precision for calibration and inspection tasks.
  • Made from durable steel for long-lasting performance.
  • 0.8 in size suitable for various measurement applications.
  • Conforms to ASME standards for reliable quality.
  • Includes a certificate of inspection for traceability and quality assurance.
  • Manufactured in Japan, known for precision engineering.