Learn More
MSC Mitutoyo Gage Block, Square, Steel, 0.1004 in, Grade 0
Catalog No. 5000533269
Encompass
This square steel gage block is designed for precision measurement, featuring an accuracy grade of 0. It includes a certificate of inspection, ensuring its reliability and adherence to strict standards for accurate applications.
  • Square shape for stable placement
  • Constructed from durable steel material
  • 0.1004 inch size for precise measurements
  • Accuracy grade 0 ensures high precision
  • Includes a certificate of inspection for guaranteed quality
  • Complies with ASME standards
  • Manufactured in Japan
Catalog No. 50-005-33269 Supplier MSC Supplier No. 08240376
This square steel gage block is designed for precision measurement, featuring an accuracy grade of 0. It includes a certificate of inspection, ensuring its reliability and adherence to strict standards for accurate applications.
  • Square shape for stable placement
  • Constructed from durable steel material
  • 0.1004 inch size for precise measurements
  • Accuracy grade 0 ensures high precision
  • Includes a certificate of inspection for guaranteed quality
  • Complies with ASME standards
  • Manufactured in Japan