Learn More
MSC Mitutoyo Gage Block, Square, Steel, 0.107 in, Grade 0
Catalog No. 5000512239
Encompass
This gage block is a precision measurement tool designed for high accuracy applications. It features a square shape and is constructed from durable steel, making it suitable for rigorous inspection tasks. The block has an overall length of 0.107 inches and conforms to Grade 0 accuracy standards, ensuring reliable performance.
  • Includes Certificate of Inspection
  • Square shape
  • Grade 0 accuracy
  • Made from steel
  • 0.107 in overall length
  • Meets ASME standards
Catalog No. 50-005-12239 Supplier MSC Supplier No. 41824889
This gage block is a precision measurement tool designed for high accuracy applications. It features a square shape and is constructed from durable steel, making it suitable for rigorous inspection tasks. The block has an overall length of 0.107 inches and conforms to Grade 0 accuracy standards, ensuring reliable performance.
  • Includes Certificate of Inspection
  • Square shape
  • Grade 0 accuracy
  • Made from steel
  • 0.107 in overall length
  • Meets ASME standards