Learn More
MSC Mitutoyo Gage Block, Square, Steel, 0.108 in, Grade 0
Catalog No. 5000534557
Encompass
This square steel gage block is designed for precise measurement applications. It features an accuracy grade of 0 and comes with a certificate of inspection, ensuring high reliability for critical tasks.
  • Square shape for stable placement and measurement
  • Grade 0 accuracy ensures high precision
  • Constructed from durable steel material
  • Size of 0.108 inches for specific measurement requirements
  • Complies with ASME standards
  • Manufactured in Japan
  • Includes a certificate of inspection for verified quality
Catalog No. 50-005-34557 Supplier MSC Supplier No. 88609557
This square steel gage block is designed for precise measurement applications. It features an accuracy grade of 0 and comes with a certificate of inspection, ensuring high reliability for critical tasks.
  • Square shape for stable placement and measurement
  • Grade 0 accuracy ensures high precision
  • Constructed from durable steel material
  • Size of 0.108 inches for specific measurement requirements
  • Complies with ASME standards
  • Manufactured in Japan
  • Includes a certificate of inspection for verified quality