Learn More
MSC Mitutoyo Gage Block, Square, Steel, 0.113 in, Grade 0
Catalog No. 5000531300
Encompass
This square steel gage block is designed for precision measurement and calibration. It features a 0.113 inch size and meets Grade 0 accuracy standards, making it suitable for demanding applications requiring high precision.
  • Square shape for versatile use
  • Grade 0 accuracy for high precision applications
  • Constructed from durable steel
  • Conforms to ASME standards
  • Includes a Certificate of Inspection for verified accuracy
Catalog No. 50-005-31300 Supplier MSC Supplier No. 84302199
This square steel gage block is designed for precision measurement and calibration. It features a 0.113 inch size and meets Grade 0 accuracy standards, making it suitable for demanding applications requiring high precision.
  • Square shape for versatile use
  • Grade 0 accuracy for high precision applications
  • Constructed from durable steel
  • Conforms to ASME standards
  • Includes a Certificate of Inspection for verified accuracy