Learn More
MSC Mitutoyo Gage Block, Square, Steel, 0.132 inch, Grade 0
Catalog No. 5000511105
Encompass
This gage block is an essential tool for precision measurement and calibration. Manufactured from durable steel and adhering to ASME standards, it ensures reliable and accurate performance. It includes a Certificate of Inspection, providing assurance of its quality and precision. Ideal for various industrial applications.
  • Essential tool for precision measurement and calibration
  • Manufactured from durable steel
  • Adheres to ASME standards
  • Ensures reliable and accurate performance
  • Includes a certificate of inspection
  • Ideal for various industrial applications
Catalog No. 50-005-11105 Supplier MSC Supplier No. 01809763
This gage block is an essential tool for precision measurement and calibration. Manufactured from durable steel and adhering to ASME standards, it ensures reliable and accurate performance. It includes a Certificate of Inspection, providing assurance of its quality and precision. Ideal for various industrial applications.
  • Essential tool for precision measurement and calibration
  • Manufactured from durable steel
  • Adheres to ASME standards
  • Ensures reliable and accurate performance
  • Includes a certificate of inspection
  • Ideal for various industrial applications