Learn More
MSC Mitutoyo Gage Block, Square, Steel, 0.45 inch, Grade 0
Catalog No. 5000530249
Encompass
This Mitutoyo Gage Block is a precision measuring tool with an accuracy grade of 0. It is constructed from steel in a square shape, with a size of 0.45 " This gage block is designed for accurate measurements and includes a Certificate of Inspection.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Square shape
  • Steel material
  • 0.45 inch size
  • Meets ASME standards
  • Manufactured in Japan
Catalog No. 50-005-30249 Supplier MSC Supplier No. 88609680
This Mitutoyo Gage Block is a precision measuring tool with an accuracy grade of 0. It is constructed from steel in a square shape, with a size of 0.45 " This gage block is designed for accurate measurements and includes a Certificate of Inspection.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Square shape
  • Steel material
  • 0.45 inch size
  • Meets ASME standards
  • Manufactured in Japan