Learn More
MSC Mitutoyo Gage Block, Steel, 0.112 inch, Grade 0, ASME 41111600
Catalog No. 5000511859
Encompass
This square steel gage block, with an overall length of 0.112 inch and an accuracy grade of 0, conforms to ASME 41111600 standards. It is ideal for precision measurement applications requiring high accuracy and reliability.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Made from steel
  • Overall length of 0.112 inch
  • Conforms to ASME 41111600 standards
  • Square shape
Catalog No. 50-005-11859 Supplier MSC Supplier No. 76633254
This square steel gage block, with an overall length of 0.112 inch and an accuracy grade of 0, conforms to ASME 41111600 standards. It is ideal for precision measurement applications requiring high accuracy and reliability.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Made from steel
  • Overall length of 0.112 inch
  • Conforms to ASME 41111600 standards
  • Square shape