Learn More
MSC Mitutoyo Gage Block, Steel, 0.139 Inch, Grade 0
Catalog No. 5000528589
Encompass
This gage block is a precision measuring tool designed for accuracy and durability in demanding industrial and laboratory environments. Made from high-quality steel, it provides a reliable standard for calibration and inspection, ensuring consistent measurements. Each block meets stringent ASME standards and includes a certificate of inspection for verified performance. Its robust construction and precise dimensions make it an essential component for maintaining measurement integrity across various applications.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Conforms to ASME standards
  • Made of steel
  • Rectangular shape
  • Size 0.139 inch
Catalog No. 50-005-28589 Supplier MSC Supplier No. 32882250
This gage block is a precision measuring tool designed for accuracy and durability in demanding industrial and laboratory environments. Made from high-quality steel, it provides a reliable standard for calibration and inspection, ensuring consistent measurements. Each block meets stringent ASME standards and includes a certificate of inspection for verified performance. Its robust construction and precise dimensions make it an essential component for maintaining measurement integrity across various applications.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Conforms to ASME standards
  • Made of steel
  • Rectangular shape
  • Size 0.139 inch