Learn More
MSC Mitutoyo Gage Block, Steel, Rectangle, 0.123 in, Grade 0
Catalog No. 5000514856
Encompass
This precision gage block is designed as a primary standard for dimensional quality control. Constructed from high-carbon, high-chrome steel, it offers high hardness, wear resistance, and excellent dimensional stability through advanced heat treatment. Its rectangular shape and optimal surface finish ensure accurate wringing. This Grade 0 gage block is ideal for use in inspection areas to verify the accuracy of plug and snap gages and for setting electronic measuring devices, adhering to ASME standards.
  • Identification number etched in block for traceability.
  • Rectangular base for accurate wringing.
  • High-carbon, high-chrome steel construction for hardness and wear resistance.
  • Grade 0 accuracy for verifying plug and snap gages.
  • Supplied with a Certificate of Inspection.
  • Adheres to ASME standards.
Catalog No. 50-005-14856 Supplier MSC Supplier No. 00396234
This precision gage block is designed as a primary standard for dimensional quality control. Constructed from high-carbon, high-chrome steel, it offers high hardness, wear resistance, and excellent dimensional stability through advanced heat treatment. Its rectangular shape and optimal surface finish ensure accurate wringing. This Grade 0 gage block is ideal for use in inspection areas to verify the accuracy of plug and snap gages and for setting electronic measuring devices, adhering to ASME standards.
  • Identification number etched in block for traceability.
  • Rectangular base for accurate wringing.
  • High-carbon, high-chrome steel construction for hardness and wear resistance.
  • Grade 0 accuracy for verifying plug and snap gages.
  • Supplied with a Certificate of Inspection.
  • Adheres to ASME standards.