Learn More
MSC Mitutoyo Gage Block, Steel, Rectangular, 0.144 inch, Grade 0
Catalog No. 5000531196
Encompass
This individual rectangular steel gage block with a nominal length of 0.144 inches is designed for high-precision applications. It features Grade 0 accuracy, making it suitable for use within inspection areas to verify the accuracy of plug and snap gages, and for setting electronic measuring devices.
  • Manufactured from high-carbon, high-chrome steel for excellent durability and dimensional stability.
  • Undergoes heat treatment technology to achieve high hardness and reduce long-term deterioration.
  • Guarantees high accuracy, furnished with a Certificate of Inspection.
  • Features optimal flatness and surface roughness, ensuring effective wringing.
  • Meets or exceeds all known specifications, including ASME B89.1.9-2002 standards for length deviations and tolerances.
  • Optional A2LA calibration is available.
Catalog No. 50-005-31196 Supplier MSC Supplier No. 86283207
This individual rectangular steel gage block with a nominal length of 0.144 inches is designed for high-precision applications. It features Grade 0 accuracy, making it suitable for use within inspection areas to verify the accuracy of plug and snap gages, and for setting electronic measuring devices.
  • Manufactured from high-carbon, high-chrome steel for excellent durability and dimensional stability.
  • Undergoes heat treatment technology to achieve high hardness and reduce long-term deterioration.
  • Guarantees high accuracy, furnished with a Certificate of Inspection.
  • Features optimal flatness and surface roughness, ensuring effective wringing.
  • Meets or exceeds all known specifications, including ASME B89.1.9-2002 standards for length deviations and tolerances.
  • Optional A2LA calibration is available.