Learn More
MSC Mitutoyo Gear Tooth Micrometer Anvil, Interchangeable Ball, 3.0 mm Diameter
Catalog No. 5000531324
Encompass
This interchangeable ball anvil-spindle tip is designed for use with gear tooth micrometers. It allows for precise measurement of the over-pin diameter of gears with its ball-tipped measuring faces, which feature precision steel (carbide) construction. This set is compatible with both electronic and metric series gear tooth micrometers.
  • Measures over-pin diameter of gears
  • Features precision steel (carbide) ball-tipped measuring face
  • Is interchangeable for various gear modules
  • Compatible with electronic and metric series gear tooth micrometers
Catalog No. 50-005-31324 Supplier MSC Supplier No. 88701487
This interchangeable ball anvil-spindle tip is designed for use with gear tooth micrometers. It allows for precise measurement of the over-pin diameter of gears with its ball-tipped measuring faces, which feature precision steel (carbide) construction. This set is compatible with both electronic and metric series gear tooth micrometers.
  • Measures over-pin diameter of gears
  • Features precision steel (carbide) ball-tipped measuring face
  • Is interchangeable for various gear modules
  • Compatible with electronic and metric series gear tooth micrometers