Learn More
MSC Mitutoyo Halogen Lamp, 24V, 150W
Catalog No. 5000534242
Encompass
This halogen lamp is engineered to provide precise illumination for various optical comparators and profile projectors, ensuring accurate measurements and inspections.
  • 24V, 150W output
  • Suitable for 14″ optical comparators
  • Compatible with various profile projector models
  • Features horizontal and vertical orientation
Catalog No. 50-005-34242 Supplier MSC Supplier No. 00401703
This halogen lamp is engineered to provide precise illumination for various optical comparators and profile projectors, ensuring accurate measurements and inspections.
  • 24V, 150W output
  • Suitable for 14″ optical comparators
  • Compatible with various profile projector models
  • Features horizontal and vertical orientation