Learn More
MSC Mitutoyo Holtest, 3-Point Internal Micrometer, 4-5 in, 0.0002 in Graduation
Catalog No. 5000536117
Encompass
The Mitutoyo 3-Point Internal Micrometer is a durable measurement tool designed for precisely measuring the internal diameter of bores. It features a self-centering design with three anvils, a constant-force ratchet for consistent measurements, and titanium-coated measuring pins for durability and blind hole measurement.
  • Titanium-coated measuring pins provide excellent durability and impact resistance.
  • Three-point design ensures self-centering action for stable measurements.
  • Constant-force ratchet ensures consistent measurements with minimal operator variation.
  • Measuring pins allow measurement to the bottom of blind holes.
  • Anvils are carbide-tipped for optimal durability.
  • Inspection certificate included.
  • Laser-etched scale on thimble and sleeve for easy reading.
  • Capable of measuring deep holes with an extension rod (sold separately).
Catalog No. 50-005-36117 Supplier MSC Supplier No. 79874194
The Mitutoyo 3-Point Internal Micrometer is a durable measurement tool designed for precisely measuring the internal diameter of bores. It features a self-centering design with three anvils, a constant-force ratchet for consistent measurements, and titanium-coated measuring pins for durability and blind hole measurement.
  • Titanium-coated measuring pins provide excellent durability and impact resistance.
  • Three-point design ensures self-centering action for stable measurements.
  • Constant-force ratchet ensures consistent measurements with minimal operator variation.
  • Measuring pins allow measurement to the bottom of blind holes.
  • Anvils are carbide-tipped for optimal durability.
  • Inspection certificate included.
  • Laser-etched scale on thimble and sleeve for easy reading.
  • Capable of measuring deep holes with an extension rod (sold separately).