Learn More
MSC Mitutoyo Holtest Three-Point Internal Micrometer, 0.425-0.500 in, 0.0001 in Graduation
Catalog No. 5000530205
Encompass
This three-point internal micrometer provides accurate and efficient measurements of bore inside dimensions. It features three self-centering anvils, spaced at 120° for true alignment, and Titanium Nitride-coated contact points for durability and precision. A ratchet stop mechanism ensures consistent measuring force. With a range of 0.425-0.500 inch and 0.0001 inch graduation, it offers reliable results.
  • Inch models available.
  • Optional extension rod for deep holes.
  • Optional setting rings for zero-point adjustment.
  • Laser-etched scale on thimble and sleeve for easy reading.
  • Supplied in a fitted plastic case.
  • Self-centering anvils for accurate inside dimension measurements.
  • Hardened-steel anvils spaced at 120° for true bore alignment.
  • Titanium Nitride (TiN) coated contact points for durability.
  • Ratchet stop mechanism for constant measuring force.
Catalog No. 50-005-30205 Supplier MSC Supplier No. 73597098
This three-point internal micrometer provides accurate and efficient measurements of bore inside dimensions. It features three self-centering anvils, spaced at 120° for true alignment, and Titanium Nitride-coated contact points for durability and precision. A ratchet stop mechanism ensures consistent measuring force. With a range of 0.425-0.500 inch and 0.0001 inch graduation, it offers reliable results.
  • Inch models available.
  • Optional extension rod for deep holes.
  • Optional setting rings for zero-point adjustment.
  • Laser-etched scale on thimble and sleeve for easy reading.
  • Supplied in a fitted plastic case.
  • Self-centering anvils for accurate inside dimension measurements.
  • Hardened-steel anvils spaced at 120° for true bore alignment.
  • Titanium Nitride (TiN) coated contact points for durability.
  • Ratchet stop mechanism for constant measuring force.