Learn More
MSC Mitutoyo Horizontal Dial Test Indicator, 0.0160 in Max, 0-5-0 Dial Reading, 0.000200 in Accuracy
Catalog No. 5000516663
Encompass
The Mitutoyo Horizontal Dial Test Indicator is engineered for precise measurements in various applications. It features a maximum measurement of 0.0160 in and an accuracy of 0.000200 in, making it suitable for detailed inspection tasks.
  • Dust and oil proof
  • O-ring seal
  • Carbide-tipped contact point
  • Glare-free crystal
  • No-clutch structure
  • Automatic reversal
  • High-impact resistance
  • Durable design
  • Scratch-resistant crystal
Catalog No. 50-005-16663 Supplier MSC Supplier No. 06385496
The Mitutoyo Horizontal Dial Test Indicator is engineered for precise measurements in various applications. It features a maximum measurement of 0.0160 in and an accuracy of 0.000200 in, making it suitable for detailed inspection tasks.
  • Dust and oil proof
  • O-ring seal
  • Carbide-tipped contact point
  • Glare-free crystal
  • No-clutch structure
  • Automatic reversal
  • High-impact resistance
  • Durable design
  • Scratch-resistant crystal