Learn More
MSC Mitutoyo Horizontal Dial Test Indicator, 0.14 mm Range, 10 μm Accuracy
Catalog No. 5000512071
Encompass
The Horizontal Dial Test Indicator is designed for precise measurements, featuring a maximum measurement of 0.14 mm, a 0-70-0 dial reading, and an accuracy of 10 μm. It is part of the Series 513 and includes a yellow dial with a 0.0010 mm dial graduation and a 12.80 mm contact point. The indicator is built with features such as being dust and oil proof, having an O-ring seal, a carbide-tipped contact point, and a glare-free crystal. Its construction also includes a no-clutch structure, automatic reversal, high-impact resistance, and durability with scratch-resistant properties.
  • Dust and oil proof
  • O-ring seal
  • Carbide-tipped
  • Glare-free crystal
  • No-clutch structure
  • Automatic reversal
  • High-impact resistance
  • Durable
  • Scratch-resistant
Catalog No. 50-005-12071 Supplier MSC Supplier No. 33811829
The Horizontal Dial Test Indicator is designed for precise measurements, featuring a maximum measurement of 0.14 mm, a 0-70-0 dial reading, and an accuracy of 10 μm. It is part of the Series 513 and includes a yellow dial with a 0.0010 mm dial graduation and a 12.80 mm contact point. The indicator is built with features such as being dust and oil proof, having an O-ring seal, a carbide-tipped contact point, and a glare-free crystal. Its construction also includes a no-clutch structure, automatic reversal, high-impact resistance, and durability with scratch-resistant properties.
  • Dust and oil proof
  • O-ring seal
  • Carbide-tipped
  • Glare-free crystal
  • No-clutch structure
  • Automatic reversal
  • High-impact resistance
  • Durable
  • Scratch-resistant