Learn More
MSC Mitutoyo Hub Micrometer, 3-4 in Range, 0.001 in Graduation
Catalog No. 5000529802
Encompass
This hub micrometer is designed for precise measurements of hub thickness and shoulder inside a bore. It features high-grade hardened steel on its frame, spindle, and anvil to deliver precise, repeatable thickness and shoulder measurements. The ratchet-stop mechanism provides uniform pressure on a workpiece for precise, repeatable measurements. An inch Vernier scale provides outside diameter measurements.
  • Measuring surfaces on the anvil and spindle are constructed of carbide to reduce wear.
  • Ratchet mechanism provides an audible click to ensure measurement is performed at a constant force.
  • Satin chrome finish on the sleeve and laser etched measurements on thimble for easier reading.
  • Frame has a baked enamel finish for durability and longer lifespan.
  • Includes a standard bar (except for 0-1 inch or 0-25 mm models).
  • Supplied in a fitted plastic case.
Catalog No. 50-005-29802 Supplier MSC Supplier No. 06235899
This hub micrometer is designed for precise measurements of hub thickness and shoulder inside a bore. It features high-grade hardened steel on its frame, spindle, and anvil to deliver precise, repeatable thickness and shoulder measurements. The ratchet-stop mechanism provides uniform pressure on a workpiece for precise, repeatable measurements. An inch Vernier scale provides outside diameter measurements.
  • Measuring surfaces on the anvil and spindle are constructed of carbide to reduce wear.
  • Ratchet mechanism provides an audible click to ensure measurement is performed at a constant force.
  • Satin chrome finish on the sleeve and laser etched measurements on thimble for easier reading.
  • Frame has a baked enamel finish for durability and longer lifespan.
  • Includes a standard bar (except for 0-1 inch or 0-25 mm models).
  • Supplied in a fitted plastic case.