Learn More
MSC Mitutoyo Indicator Spindle Lifting Cable, 1 in Lift, 11.81 in Cable
Catalog No. 5000530354
Encompass
This indicator spindle lifting cable is designed to securely attach to most digital indicators via a screw connection. It allows users to raise the indicator's spindle by pressing a plunger on the cable and lowers it back into position upon release. This tool is ideal for lifting the spindle to facilitate part changes during measurements, eliminating the need to directly handle the indicator.
  • Lifting cable accessory
  • Compatible with most digital indicators
  • 1 in (25.4 mm) lifting range
  • 11.81 in (300 mm) cable length
  • Includes mounting accessory
  • Without auto-stop function
  • Attaches securely via screw connection
  • Facilitates part changes during measurements
Catalog No. 50-005-30354 Supplier MSC Supplier No. 63909238
This indicator spindle lifting cable is designed to securely attach to most digital indicators via a screw connection. It allows users to raise the indicator's spindle by pressing a plunger on the cable and lowers it back into position upon release. This tool is ideal for lifting the spindle to facilitate part changes during measurements, eliminating the need to directly handle the indicator.
  • Lifting cable accessory
  • Compatible with most digital indicators
  • 1 in (25.4 mm) lifting range
  • 11.81 in (300 mm) cable length
  • Includes mounting accessory
  • Without auto-stop function
  • Attaches securely via screw connection
  • Facilitates part changes during measurements